Product Details

SNP ID
rs430206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:169426971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCAGTGCTGTGGTGGTGGGGGC[A/G]GTGGGGGGGGGGTGGCGGTTGCATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC401287 PubMed Links

Gene Details

Gene
LOC401287
Gene Name
uncharacterized LOC401287
There are no transcripts associated with this gene.

Gene
WDR27
Gene Name
WD repeat domain 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202550.1 4043 Intron NP_001189479.1
NM_182552.4 4043 Intron NP_872358.4
XM_011535682.2 4043 Intron XP_011533984.1
XM_011535684.2 4043 Intron XP_011533986.1
XM_011535685.2 4043 Intron XP_011533987.1
XM_011535687.2 4043 UTR 3 XP_011533989.1
XM_011535688.2 4043 UTR 3 XP_011533990.1
XM_011535689.2 4043 UTR 3 XP_011533991.1
XM_011535690.2 4043 Intron XP_011533992.1
XM_011535691.2 4043 Intron XP_011533993.1
XM_011535692.2 4043 Intron XP_011533994.1
XM_011535693.2 4043 Intron XP_011533995.1
XM_011535694.2 4043 Intron XP_011533996.1
XM_011535696.2 4043 Intron XP_011533998.1
XM_011535697.2 4043 Intron XP_011533999.1
XM_017010658.1 4043 Intron XP_016866147.1
XM_017010659.1 4043 Intron XP_016866148.1
XM_017010660.1 4043 Intron XP_016866149.1
XM_017010661.1 4043 UTR 3 XP_016866150.1
XM_017010662.1 4043 Intron XP_016866151.1
XM_017010663.1 4043 Intron XP_016866152.1
XM_017010664.1 4043 Intron XP_016866153.1
XM_017010665.1 4043 Intron XP_016866154.1
XM_017010666.1 4043 Intron XP_016866155.1
XM_017010667.1 4043 Intron XP_016866156.1
XM_017010668.1 4043 Intron XP_016866157.1
XM_017010669.1 4043 Intron XP_016866158.1
XM_017010670.1 4043 Intron XP_016866159.1
XM_017010671.1 4043 Intron XP_016866160.1
XM_017010672.1 4043 Intron XP_016866161.1
XM_017010673.1 4043 Intron XP_016866162.1
XM_017010674.1 4043 Intron XP_016866163.1
XM_017010675.1 4043 Intron XP_016866164.1

View Full Product Details