Product Details

SNP ID
rs35879117
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:171848126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAACTTAATGTTAGGAACTGGGA[A/G]GCCTGTGCCAGTTCATCATCTTCTC
Phenotype
MIM: 611445
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNM3 PubMed Links

Gene Details

Gene
DNM3
Gene Name
dynamin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136127.2 Intron NP_001129599.1
NM_001278252.1 Intron NP_001265181.1
NM_015569.4 Intron NP_056384.2
XM_005245079.1 Intron XP_005245136.1
XM_005245080.1 Intron XP_005245137.1
XM_006711268.2 Intron XP_006711331.1
XM_017000976.1 Intron XP_016856465.1
XM_017000977.1 Intron XP_016856466.1
XM_017000978.1 Intron XP_016856467.1
XM_017000979.1 Intron XP_016856468.1
XM_017000980.1 Intron XP_016856469.1
XM_017000981.1 Intron XP_016856470.1
XM_017000982.1 Intron XP_016856471.1
XM_017000983.1 Intron XP_016856472.1
XM_017000984.1 Intron XP_016856473.1
XM_017000985.1 Intron XP_016856474.1
XM_017000986.1 Intron XP_016856475.1
XM_017000987.1 Intron XP_016856476.1
XM_017000988.1 Intron XP_016856477.1
XM_017000989.1 Intron XP_016856478.1
XM_017000990.1 Intron XP_016856479.1
XM_017000991.1 Intron XP_016856480.1
XM_017000992.1 Intron XP_016856481.1

View Full Product Details