Product Details

SNP ID
rs2086298
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:82908176 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCAAGACCTAAGGGAAGAATATC[A/G]TAGCCAAAGAGAATAGTAAGCACAA
Phenotype
MIM: 176785
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDIAS PubMed Links

Gene Details

Gene
DDIAS
Gene Name
DNA damage induced apoptosis suppressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145018.3 Intron NP_659455.3
XM_011544835.1 Intron XP_011543137.1
XM_011544836.2 Intron XP_011543138.1
XM_011544837.1 Intron XP_011543139.1
XM_011544838.2 Intron XP_011543140.1
XM_017017352.1 Intron XP_016872841.1
XM_017017353.1 Intron XP_016872842.1
Gene
PRCP
Gene Name
prolylcarboxypeptidase
There are no transcripts associated with this gene.

View Full Product Details