Product Details

SNP ID
rs319590
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:134919098 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAATGGACTTTTAACTGCATAATTT[G/T]CTCTTAGTCATGTAGTAAATCTTCT
Phenotype
MIM: 609836
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR4461 PubMed Links

Gene Details

Gene
MIR4461
Gene Name
microRNA 4461
There are no transcripts associated with this gene.

Gene
PCBD2
Gene Name
pterin-4 alpha-carbinolamine dehydratase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032151.4 Intron NP_115527.3

View Full Product Details