Product Details

SNP ID
rs2282083
Assay Type
Validated
NCBI dbSNP Submissions
56
Location
Chr.1:229325277 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
AGCACCTCATGTATTCTGTCATCCA[C/T]GGGTTCTCTGAGGAGGAAGCCTTCA
Phenotype
MIM: 616762
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCSAP PubMed Links

Gene Details

Gene
CCSAP
Gene Name
centriole, cilia and spindle associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145257.4 849 Silent Mutation CCA,CCG P257P NP_660300.3

View Full Product Details