Product Details

SNP ID
rs6676143
Assay Type
Functionally Tested
NCBI dbSNP Submissions
37
Location
Chr.1:16201723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAACACAGCACCCAGGATGGGAG[G/T]GGAGGAGAGCGGGCGAGGGTCCAGC
Phenotype
MIM: 612496
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ARHGEF19 PubMed Links

Gene Details

Gene
ARHGEF19
Gene Name
Rho guanine nucleotide exchange factor 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011540706.2 Intron XP_011539008.1

View Full Product Details