Product Details

SNP ID
rs2658800
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:93480800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGAGTTGGGAAAGGTCTTACTGA[C/T]ACCATCCTGTTTTTATGAGGGATAC
Phenotype
MIM: 609477
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMCO4 PubMed Links

Gene Details

Gene
SMCO4
Gene Name
single-pass membrane protein with coiled-coil domains 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020179.2 Intron NP_064564.1
XM_011542907.1 Intron XP_011541209.1
XM_011542908.2 Intron XP_011541210.1
XM_011542909.1 Intron XP_011541211.1
XM_011542910.2 Intron XP_011541212.1
XM_011542911.2 Intron XP_011541213.1
XM_017018019.1 Intron XP_016873508.1
XM_017018020.1 Intron XP_016873509.1

View Full Product Details