Product Details

SNP ID
rs6778
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:52497607 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTTTTTACATGAAAGCATAAATACG[A/G]CAGGTACAAATTTTCCTTGAGTTCC
Phenotype
MIM: 608815 MIM: 608485
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EFHC1 PubMed Links

Gene Details

Gene
EFHC1
Gene Name
EF-hand domain containing 1
There are no transcripts associated with this gene.

Gene
TRAM2
Gene Name
translocation associated membrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012288.3 6727 UTR 3 NP_036420.1
XM_011515005.2 6727 UTR 3 XP_011513307.1

View Full Product Details