Product Details

SNP ID
rs2224198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52741111 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAAATCAAATCCAGTGTTGTTCT[A/G]AAGACTGTAGACCAAATTCTACTGT
Phenotype
MIM: 138360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSTA2 PubMed Links

Gene Details

Gene
GSTA2
Gene Name
glutathione S-transferase alpha 2
There are no transcripts associated with this gene.

Gene
GSTA7P
Gene Name
glutathione S-transferase alpha 7, pseudogene
There are no transcripts associated with this gene.

View Full Product Details