Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001168374.1 | 3708 | UTR 3 | NP_001161846.1 | ||
NM_001168375.1 | 3708 | UTR 3 | NP_001161847.1 | ||
NM_001168376.1 | 3708 | UTR 3 | NP_001161848.1 | ||
NM_001168377.1 | 3708 | UTR 3 | NP_001161849.1 | ||
NM_001252328.1 | 3708 | UTR 3 | NP_001239257.1 | ||
NM_014809.3 | 3708 | UTR 3 | NP_055624.2 | ||
XM_011515025.2 | 3708 | UTR 3 | XP_011513327.1 | ||
XM_011515026.2 | 3708 | UTR 3 | XP_011513328.1 | ||
XM_017011541.1 | 3708 | UTR 3 | XP_016867030.1 | ||
XM_017011542.1 | 3708 | Intron | XP_016867031.1 | ||
XM_017011543.1 | 3708 | UTR 3 | XP_016867032.1 | ||
XM_017011544.1 | 3708 | UTR 3 | XP_016867033.1 | ||
XM_017011545.1 | 3708 | UTR 3 | XP_016867034.1 | ||
XM_017011546.1 | 3708 | Intron | XP_016867035.1 | ||
XM_017011547.1 | 3708 | UTR 3 | XP_016867036.1 | ||
XM_017011548.1 | 3708 | UTR 3 | XP_016867037.1 | ||
XM_017011549.1 | 3708 | UTR 3 | XP_016867038.1 | ||
XM_017011550.1 | 3708 | Intron | XP_016867039.1 | ||
XM_017011551.1 | 3708 | UTR 3 | XP_016867040.1 | ||
XM_017011552.1 | 3708 | UTR 3 | XP_016867041.1 | ||
XM_017011553.1 | 3708 | UTR 3 | XP_016867042.1 |