Product Details

SNP ID
rs13286733
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136659739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGAGACAATGGGGAAACTGAGG[A/C]CTGTGTGGGGCACAAAGCTGGGACC
Phenotype
MIM: 608582
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EGFL7 PubMed Links

Gene Details

Gene
EGFL7
Gene Name
EGF like domain multiple 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016215.4 Intron NP_057299.1
NM_201446.2 Intron NP_958854.1
XM_006717141.3 Intron XP_006717204.1
XM_011518764.1 Intron XP_011517066.1
XM_011518765.1 Intron XP_011517067.1
XM_011518766.1 Intron XP_011517068.1
XM_011518767.1 Intron XP_011517069.1
XM_011518768.1 Intron XP_011517070.1
XM_017014795.1 Intron XP_016870284.1
XM_017014796.1 Intron XP_016870285.1
XM_017014797.1 Intron XP_016870286.1
XM_017014798.1 Intron XP_016870287.1
XM_017014799.1 Intron XP_016870288.1
Gene
HSPC324
Gene Name
uncharacterized LOC101928612
There are no transcripts associated with this gene.

View Full Product Details