Product Details

SNP ID
rs6765
Assay Type
Functionally tested
NCBI dbSNP Submissions
41
Location
Chr.1:109096568 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCTTGTGTTGTTACATTTTGAGA[G/T]GTTTTCATACCAGAATGTACCAAAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C1orf194 PubMed Links

Gene Details

Gene
C1orf194
Gene Name
chromosome 1 open reading frame 194
There are no transcripts associated with this gene.

Gene
SCARNA2
Gene Name
small Cajal body-specific RNA 2
There are no transcripts associated with this gene.

Gene
TMEM167B
Gene Name
transmembrane protein 167B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322248.1 2386 UTR 3 NP_001309177.1
NM_020141.3 2386 UTR 3 NP_064526.1

View Full Product Details