Product Details

SNP ID
rs10071088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:58970625 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTATGTAAACTGACATCAACCCA[A/C]CTTTGCGTTTAAATAGATGGGTTTA
Phenotype
MIM: 600129
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PDE4D PubMed Links

Gene Details

Gene
PDE4D
Gene Name
phosphodiesterase 4D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104631.1 6286 UTR 3 NP_001098101.1
NM_001165899.1 6286 UTR 3 NP_001159371.1
NM_001197218.1 6286 UTR 3 NP_001184147.1
NM_001197219.1 6286 UTR 3 NP_001184148.1
NM_001197220.1 6286 UTR 3 NP_001184149.1
NM_001197221.1 6286 UTR 3 NP_001184150.1
NM_001197222.1 6286 UTR 3 NP_001184151.1
NM_001197223.1 6286 UTR 3 NP_001184152.1
NM_006203.4 6286 UTR 3 NP_006194.2
XM_005248537.3 6286 UTR 3 XP_005248594.1
XM_005248538.4 6286 UTR 3 XP_005248595.1
XM_011543469.1 6286 UTR 3 XP_011541771.1
XM_011543470.2 6286 UTR 3 XP_011541772.1
XM_011543471.1 6286 UTR 3 XP_011541773.1
XM_011543473.1 6286 UTR 3 XP_011541775.1
XM_011543477.2 6286 UTR 3 XP_011541779.1
XM_017009565.1 6286 UTR 3 XP_016865054.1
XM_017009566.1 6286 UTR 3 XP_016865055.1
XM_017009567.1 6286 UTR 3 XP_016865056.1
XM_017009568.1 6286 UTR 3 XP_016865057.1
XM_017009569.1 6286 UTR 3 XP_016865058.1
XM_017009570.1 6286 UTR 3 XP_016865059.1
XM_017009571.1 6286 UTR 3 XP_016865060.1

View Full Product Details