Product Details

SNP ID
rs12459002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:62733478 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCATGTGACTTTATTGTGATATTC[A/G]CCCTATAACAGTGGTCTGGAACTGA
Phenotype
MIM: 609396
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PHLPP1 PubMed Links

Gene Details

Gene
PHLPP1
Gene Name
PH domain and leucine rich repeat protein phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194449.3 Intron NP_919431.2
XM_011525886.1 Intron XP_011524188.1

View Full Product Details