Product Details
- SNP ID
-
rs12562208
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
12
- Location
-
Chr.1:33145203 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTTTTGGACAGGTTTGTGGTGTGG[C/T]ACTTGCCAGCATGGGATATGGGACC
- Phenotype
-
MIM: 608353
MIM: 616755
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AZIN2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80169825] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AZIN2
- Gene Name
- antizyme inhibitor 2
- Gene
- TRIM62
- Gene Name
- tripartite motif containing 62
View Full Product Details