Product Details

SNP ID
rs3825016
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64591814 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCCGGGCCCCAACCAGAGGCCCCA[C/T]CAGTGCCGCCGCTTCCGCCAGCCAC
Phenotype
MIM: 607096
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC22A12 PubMed Links

Gene Details

Gene
SLC22A12
Gene Name
solute carrier family 22 member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276326.1 1005 Silent Mutation CAC,CAT H86H NP_001263255.1
NM_001276327.1 1005 Silent Mutation CAC,CAT H86H NP_001263256.1
NM_144585.3 1005 Silent Mutation CAC,CAT H86H NP_653186.2
NM_153378.2 1005 UTR 5 NP_700357.1
XM_006718430.3 1005 Silent Mutation CAC,CAT H86H XP_006718493.1
XM_006718431.3 1005 Silent Mutation CAC,CAT H51H XP_006718494.1

View Full Product Details