Product Details

SNP ID
rs2373459
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:101480178 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTGACAGTCAACATGGATGATCA[C/T]ATGCTGTTAGGAAGGAAATAACCCT
Phenotype
MIM: 612568
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPIC PubMed Links

Gene Details

Gene
SPIC
Gene Name
Spi-C transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005268646.3 Intron XP_005268703.1
XM_006719239.3 Intron XP_006719302.1

View Full Product Details