Product Details

SNP ID
rs656721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:133399265 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATGGAGAAGCAGAGGTGTAGGGA[A/G]TGGCCGGAACATGTGTGGGATCTGA
Phenotype
MIM: 603212
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BFSP2 PubMed Links

Gene Details

Gene
BFSP2
Gene Name
beaded filament structural protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003571.3 1384 Intron NP_003562.1
XM_017007315.1 1384 UTR 5 XP_016862804.1
Gene
TMEM108
Gene Name
transmembrane protein 108
There are no transcripts associated with this gene.

View Full Product Details