Product Details
- SNP ID
-
rs11081866
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:34503775 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCCTGTTATTTACTTGAGCATTTA[C/T]AGAATTCCACTTCTATATGTATAGT
- Phenotype
-
MIM: 601239
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DTNA
PubMed Links
Gene Details
- Gene
- DTNA
- Gene Name
- dystrobrevin alpha
View Full Product Details