Product Details
- SNP ID
-
hCV3120673
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:179957547 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAATCACTATTATGCACCCAGTGC[C/T]TAGCATATGGTAACATAGTCAATAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CEP350
PubMed Links
Gene Details
- Gene
- CEP350
- Gene Name
- centrosomal protein 350
There are no transcripts associated with this gene.
- Gene
- LOC101928933
- Gene Name
- uncharacterized LOC101928933
There are no transcripts associated with this gene.
View Full Product Details