Product Details

SNP ID
rs34532055
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:51241499 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAATTCCAGTAATATTAAATACAG[A/C]AATTGGTTTACACTAAGTGTTAAGT
Phenotype
MIM: 612598
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RNF11 PubMed Links

Gene Details

Gene
RNF11
Gene Name
ring finger protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014372.4 Intron NP_055187.1

View Full Product Details