Product Details

SNP ID
rs6968352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:130671124 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTCCCTACCCCAATCCCAGAGAC[C/T]TTTGTACCTCTAAGACACACAGACT
Phenotype
MIM: 604355
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COPG2 PubMed Links

Gene Details

Gene
COPG2
Gene Name
coatomer protein complex subunit gamma 2
There are no transcripts associated with this gene.

Gene
TSGA13
Gene Name
testis specific 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304968.1 Intron NP_001291897.1
NM_052933.3 Intron NP_443165.1

View Full Product Details