Product Details

SNP ID
rs11711287
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:10284412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAGGGGCATGGCTGTCTGAGGGT[A/G]GAGGGGTGAGTAGGGGGCCTGCAGT
Phenotype
MIM: 605353
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GHRL PubMed Links

Gene Details

Gene
GHRL
Gene Name
ghrelin/obestatin prepropeptide
There are no transcripts associated with this gene.

Gene
GHRLOS
Gene Name
ghrelin opposite strand/antisense RNA
There are no transcripts associated with this gene.

Gene
LINC00852
Gene Name
long intergenic non-protein coding RNA 852
There are no transcripts associated with this gene.

Gene
TATDN2
Gene Name
TatD DNase domain containing 2
There are no transcripts associated with this gene.

View Full Product Details