Product Details

SNP ID
rs175634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50606703 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCAGACCTAGCAGAACTGCAGTGG[C/T]GGCCCTCAAACCAGCACCCCCTCTT
Phenotype
MIM: 602441 MIM: 602130
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CISH PubMed Links

Gene Details

Gene
CISH
Gene Name
cytokine inducible SH2 containing protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013324.5 4684 UTR 3 NP_037456.5
NM_145071.2 4684 UTR 3 NP_659508.1
XM_011533329.2 4684 UTR 3 XP_011531631.1
Gene
MAPKAPK3
Gene Name
mitogen-activated protein kinase-activated protein kinase 3
There are no transcripts associated with this gene.

View Full Product Details