Product Details
- SNP ID
-
rs12445971
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:21262958 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- CTCACAAAAATCCCGAAACAGAGAC[A/G]TCACTTGGTGCTTATTCACACCTCA
- Phenotype
-
MIM: 123740
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CRYM
PubMed Links
Gene Details
- Gene
- CRYM
- Gene Name
- crystallin mu
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001888.4 |
|
Intron |
|
|
NP_001879.1 |
View Full Product Details