Product Details
- SNP ID
-
rs9470131
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:35843886 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTAATAGAAAAACCTCTTACAGTG[C/T]TCGAGAGTGTCTTAGGAAATGATAT
- Phenotype
-
MIM: 601939
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SRPK1
PubMed Links
Gene Details
- Gene
- SRPK1
- Gene Name
- SRSF protein kinase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003137.4 |
|
Intron |
|
|
NP_003128.3 |
View Full Product Details