Product Details

SNP ID
rs929762
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:170783816 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGGACCCAGAAGACCGTGCCTTGCC[C/T]GGAAGTCCTGCCTGTAGGCCTGAAG
Phenotype
MIM: 602729
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GABRP PubMed Links

Gene Details

Gene
GABRP
Gene Name
gamma-aminobutyric acid type A receptor pi subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291985.1 98 UTR 5 NP_001278914.1
NM_014211.2 98 UTR 5 NP_055026.1
XM_005265872.2 98 Intron XP_005265929.1

View Full Product Details