Product Details
- SNP ID
-
rs33554
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:112977039 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CCCCGTCGGGTTTTCTCAGTTTCGC[C/G]GACCCCCAGAGGCCTCTGGGTCTTC
- Phenotype
-
MIM: 609844
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DCP2
PubMed Links
Gene Details
- Gene
- DCP2
- Gene Name
- decapping mRNA 2
View Full Product Details