Product Details
- SNP ID
-
rs11747607
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:154452729 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGAAGGCTGGGCCTAGCAAAGAGC[G/T]ATTTGAGGATAACAGCCCTCCTTTG
- Phenotype
-
MIM: 610398
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SAP30L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76319463] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SAP30L
- Gene Name
- SAP30 like
- Gene
- SAP30L-AS1
- Gene Name
- SAP30L antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
View Full Product Details