Product Details

SNP ID
rs35618752
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63845492 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGATACTTCTGTGATGCTTTTC[C/T]GCCTGTATGTGAAACTTCAATAAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C20orf181 PubMed Links

Gene Details

Gene
C20orf181
Gene Name
chromosome 20 open reading frame 181
There are no transcripts associated with this gene.

View Full Product Details