Product Details
- SNP ID
-
rs11811007
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
33
- Location
-
Chr.1:6524597 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTCCCACAGGCAATAATGGAGCTT[C/T]CCCAAGAGCTGGTCTGCCAGTCCTT
- Phenotype
-
MIM: 611101
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NOL9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76388729] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NOL9
- Gene Name
- nucleolar protein 9
- Gene
- PLEKHG5
- Gene Name
- pleckstrin homology and RhoGEF domain containing G5
There are no transcripts associated with this gene.
View Full Product Details