Product Details
- SNP ID
-
rs12553716
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:214142 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCACCTTGAAGCGAACACGGGCTC[C/T]TCCACTCCCGTCAACACTGGGACTT
- Phenotype
-
MIM: 611432
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C9orf66
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116662320] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C9orf66
- Gene Name
- chromosome 9 open reading frame 66
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152569.2 |
1752 |
UTR 3 |
|
|
NP_689782.2 |
- Gene
- DOCK8
- Gene Name
- dedicator of cytokinesis 8
View Full Product Details