Product Details

SNP ID
rs1111930
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:126308155 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAGAATATTCCATGCCAAAAGCCAG[A/G]TGTGACATGTGCATTTTTTCTCAAG
Phenotype
MIM: 610534
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCPS PubMed Links

Gene Details

Gene
DCPS
Gene Name
decapping enzyme, scavenger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014026.4 Intron NP_054745.1
XM_011542778.2 Intron XP_011541080.1
XM_011542780.2 Intron XP_011541082.1
XM_017017612.1 Intron XP_016873101.1

View Full Product Details