Product Details
- SNP ID
-
rs903164
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:2036208 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TGCTTGGAGACACAAGGTCCCTCCT[A/G]GTTTCTGGGGTGGCCTCTGCCTTCC
- Phenotype
-
MIM: 603527
MIM: 607896
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DPH1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3752963] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DPH1
- Gene Name
- diphthamide biosynthesis 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001383.3 |
|
Intron |
|
|
NP_001374.3 |
- Gene
- OVCA2
- Gene Name
- ovarian tumor suppressor candidate 2
There are no transcripts associated with this gene.
View Full Product Details