Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001123066.3 | Intron | NP_001116538.2 | |||
NM_001123067.3 | Intron | NP_001116539.1 | |||
NM_001203251.1 | Intron | NP_001190180.1 | |||
NM_001203252.1 | Intron | NP_001190181.1 | |||
NM_005910.5 | Intron | NP_005901.2 | |||
NM_016834.4 | Intron | NP_058518.1 | |||
NM_016835.4 | Intron | NP_058519.3 | |||
NM_016841.4 | Intron | NP_058525.1 | |||
XM_005257362.4 | Intron | XP_005257419.1 | |||
XM_005257364.4 | Intron | XP_005257421.1 | |||
XM_005257365.4 | Intron | XP_005257422.1 | |||
XM_005257366.3 | Intron | XP_005257423.1 | |||
XM_005257367.4 | Intron | XP_005257424.1 | |||
XM_005257368.4 | Intron | XP_005257425.1 | |||
XM_005257369.4 | Intron | XP_005257426.1 | |||
XM_005257370.4 | Intron | XP_005257427.1 | |||
XM_005257371.4 | Intron | XP_005257428.1 |