Product Details
- SNP ID
-
rs1149714
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:75288321 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAGAGTAGAGGAAGTGTGCCTGGG[C/T]CTGGGGCCTCAGAGCTGCGAGATGA
- Phenotype
-
MIM: 613902
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF503
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114083421] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF503
- Gene Name
- zinc finger protein 503
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032772.5 |
|
Intron |
|
|
NP_116161.2 |
- Gene
- ZNF503-AS1
- Gene Name
- ZNF503 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details