Product Details
- SNP ID
-
rs495869
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:125789528 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACCAGCGTAAGTTAGAGGGTCAGGG[A/C]AGGTGTTGTAAGATTCTTAGAATTC
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PATE3
PubMed Links
Gene Details
- Gene
- PATE3
- Gene Name
- prostate and testis expressed 3
View Full Product Details