Product Details

SNP ID
rs149592
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3674828 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGGGGAAACCTTCTCTGGCTACT[A/C]CTCGCTGACCTCTCCGACACCCCAT
Phenotype
MIM: 602836 MIM: 616484
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EMC6 PubMed Links

Gene Details

Gene
EMC6
Gene Name
ER membrane protein complex subunit 6
There are no transcripts associated with this gene.

Gene
P2RX5
Gene Name
purinergic receptor P2X 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204519.1 Intron NP_001191448.1
NM_001204520.1 Intron NP_001191449.1
NM_002561.3 Intron NP_002552.2
NM_175080.2 Intron NP_778255.1
Gene
P2RX5-TAX1BP3
Gene Name
P2RX5-TAX1BP3 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
TAX1BP3
Gene Name
Tax1 binding protein 3
There are no transcripts associated with this gene.

View Full Product Details