Product Details

SNP ID
rs3135021
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:33077781 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCACACCCCTCCTAGGACACACCTA[A/G]AAGGTACTGACATCATGTCACCTCC
Phenotype
MIM: 142880 MIM: 142858
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HLA-DPA1 PubMed Links
Additional Information
For this assay, SNP(s) [rs28449420] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
HLA-DPA1
Gene Name
major histocompatibility complex, class II, DP alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242524.1 Intron NP_001229453.1
NM_001242525.1 Intron NP_001229454.1
NM_033554.3 Intron NP_291032.2
Gene
HLA-DPB1
Gene Name
major histocompatibility complex, class II, DP beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002121.5 Intron NP_002112.3

View Full Product Details