Product Details

SNP ID
rs303465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:88761090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTCTGAATCTTTAGATATTTTG[G/T]CAAATTGTAGGCAAACAAAGACTTA
Phenotype
MIM: 613922 MIM: 613924
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LIPK PubMed Links

Gene Details

Gene
LIPK
Gene Name
lipase family member K
There are no transcripts associated with this gene.

Gene
LIPN
Gene Name
lipase family member N
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102469.1 Intron NP_001095939.1
XM_005270049.3 Intron XP_005270106.1
XM_011540083.2 Intron XP_011538385.1
XM_011540084.2 Intron XP_011538386.1
XM_011540085.2 Intron XP_011538387.1
XM_011540086.2 Intron XP_011538388.1
XM_017016549.1 Intron XP_016872038.1

View Full Product Details