Product Details
- SNP ID
-
rs6471912
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:61072140 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCTCCATGTGATCTGTCTGCTGCA[C/T]GTCTCGGTTCATCTCCAGCTTTCCT
- Phenotype
-
MIM: 611292
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CLVS1
PubMed Links
Gene Details
- Gene
- CLVS1
- Gene Name
- clavesin 1
View Full Product Details