Product Details

SNP ID
rs34817489
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89617030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGCAGGCCCTGGAGCAGAAGGG[A/G]AAGGGAGGGGTGGGGTGATGGGACT
Phenotype
MIM: 179780
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPEP1 PubMed Links

Gene Details

Gene
DPEP1
Gene Name
dipeptidase 1 (renal)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128141.2 Intron NP_001121613.1
NM_004413.3 Intron NP_004404.1
XM_005256285.4 Intron XP_005256342.1
XM_005256286.3 Intron XP_005256343.1
XM_011522925.2 Intron XP_011521227.1
XM_017023009.1 Intron XP_016878498.1

View Full Product Details