Product Details
- SNP ID
-
rs34013592
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:56920636 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TAGGAGGCAGCACCTATACCTGCTT[C/T]ATACAATTCAGTTATAATCCTTACT
- Phenotype
-
MIM: 600480
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC145783
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75578967] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC145783
- Gene Name
- uncharacterized LOC145783
There are no transcripts associated with this gene.
- Gene
- TCF12
- Gene Name
- transcription factor 12
View Full Product Details