Product Details

SNP ID
rs2164885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52697253 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATAAGTCTTATTCATTTTCAAAA[C/T]GGATTAGACTATCCAAGTTTCCCCA
Phenotype
MIM: 608011 MIM: 610358
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLT8D1 PubMed Links

Gene Details

Gene
GLT8D1
Gene Name
glycosyltransferase 8 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010983.2 Intron NP_001010983.1
NM_001278280.1 Intron NP_001265209.1
NM_001278281.1 Intron NP_001265210.1
NM_018446.3 Intron NP_060916.1
NM_152932.2 Intron NP_690909.1
XM_017006857.1 Intron XP_016862346.1
XM_017006858.1 Intron XP_016862347.1
XM_017006859.1 Intron XP_016862348.1
Gene
GNL3
Gene Name
G protein nucleolar 3
There are no transcripts associated with this gene.

Gene
SNORD136
Gene Name
small nucleolar RNA, C/D box 136
There are no transcripts associated with this gene.

Gene
SNORD19
Gene Name
small nucleolar RNA, C/D box 19
There are no transcripts associated with this gene.

Gene
SNORD19B
Gene Name
small nucleolar RNA, C/D box 19B
There are no transcripts associated with this gene.

Gene
SNORD69
Gene Name
small nucleolar RNA, C/D box 69
There are no transcripts associated with this gene.

Gene
SPCS1
Gene Name
signal peptidase complex subunit 1
There are no transcripts associated with this gene.

View Full Product Details