Product Details

SNP ID
rs8032846
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.15:41313455 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAAAGTATGTGTCAAAAAAAGCCTG[C/G]CTTCTTAATGATTTTCATTTCAAGA
Phenotype
MIM: 606020
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OIP5 PubMed Links

Gene Details

Gene
OIP5
Gene Name
Opa interacting protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317860.1 Intron NP_001304789.1
NM_007280.1 Intron NP_009211.1

View Full Product Details