Product Details

SNP ID
rs13039047
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:60049835 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTATTTTAGGTTGTTGTGACTTCC[A/C]ATCAAGAGGTCCTGACTAATATATG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C20orf197 PubMed Links

Gene Details

Gene
C20orf197
Gene Name
chromosome 20 open reading frame 197
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302813.1 Intron NP_001289742.1
NM_001302815.1 Intron NP_001289744.1
NM_173644.2 Intron NP_775915.1
XM_017027828.1 Intron XP_016883317.1
XM_017027829.1 Intron XP_016883318.1
XM_017027830.1 Intron XP_016883319.1
XM_017027831.1 Intron XP_016883320.1
XM_017027832.1 Intron XP_016883321.1

View Full Product Details