Product Details

SNP ID
rs2289389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:49130688 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTCTTTGACAGTGGCAAAGAAAG[G/T]AATAGAATCCAGTTGGCAGAAAGAA
Phenotype
MIM: 604508
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
COPS2 PubMed Links

Gene Details

Gene
COPS2
Gene Name
COP9 signalosome subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143887.1 Intron NP_001137359.1
NM_004236.3 Intron NP_004227.1

View Full Product Details