Product Details

SNP ID
rs627911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:126202793 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTAACAGCTGTTAGACAGCATGC[C/T]GCTCCCCGCTACTGCCTTACACCCA
Phenotype
MIM: 616587
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM118B PubMed Links

Gene Details

Gene
FAM118B
Gene Name
family with sequence similarity 118 member B
There are no transcripts associated with this gene.

Gene
RPUSD4
Gene Name
RNA pseudouridylate synthase domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144827.1 1810 UTR 3 NP_001138299.1
NM_032795.2 1810 UTR 3 NP_116184.2
XM_011543039.2 1810 UTR 3 XP_011541341.1

View Full Product Details