Product Details

SNP ID
rs12589375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:92118036 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTGCATCTGAAACAAGAAGATGA[C/T]AGACATAGATGTAATAGATGTTAAA
Phenotype
MIM: 606028 MIM: 603837
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPSF2 PubMed Links

Gene Details

Gene
CPSF2
Gene Name
cleavage and polyadenylation specific factor 2
There are no transcripts associated with this gene.

Gene
NDUFB1
Gene Name
NADH:ubiquinone oxidoreductase subunit B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004545.3 Intron NP_004536.2

View Full Product Details