Product Details

SNP ID
rs3781560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:116622009 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAATACCTCACTGCCCAATTTGG[C/T]CCCCCAAGTCCTGCTGTAGGAGACA
Phenotype
MIM: 604423
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PNLIPRP2 PubMed Links

Gene Details

Gene
PNLIPRP2
Gene Name
pancreatic lipase related protein 2 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005396.4 Intron NP_005387.2
XM_017016339.1 Intron XP_016871828.1

View Full Product Details